Learn More
Grainger Temperature/Humdity Meter, 0 to 100 per Rel Hum Rng
Supplier:  Grainger 4GE36
Encompass_Preferred
The Extech by FLIR hygro-thermometer psychrometer is ideal for field applications. Simultaneously displays relative humidity (10 to 90 percent RH), air temperature and dew point or wet bulb. Calculates T1-T2 differential (air temperature minus external probe temperature) using optional probe (TP890), and T2 minus dew point. Compact meter is well-suited for field applications. Unique sensor cap closes to protect to protect meter during storage. Advanced features include Data Hold, selectable Auto Power Off, and MAX/MIN/Average reading.
Catalog No. NC1710649
Explore available promotions