Learn More
Cole-Parmer Extech EA30-NIST Easyview Wide-Range Light Meter with NIST-Traceable Certificate
Supplier:  Cole-Parmer 0158866
Encompass_Preferred
This meter measures light intensity up to 400,000 lux in five ranges and 40,000 footcandles in four ranges. Unit is color and cosine corrected. Large LCD simultaneously displays readings numerically and in a bar graph format. Meter utilizes a precision silicon photodiode sensor to obtain readings.
Catalog No. NC2005700