Learn More
Cole-Parmer Extech EX800 Waterproof Pocket Water Quality Tester
Supplier:  Cole-Parmer 9959820
Encompass
Easy-to-use water quality pocket tester kit features everything needed for water testing and measurement. Reliable pocket tester with interchangeable sensors for pH and chlorine allows you to be flexible in the field. Bundled calibration solutions and sample cups give you all the necessary accessories to begin taking readings. Additional tester features include a built-in smart chip that simplifies calibration and error diagnosis. LCD simultaneously shows ppm or pH, and °F/°C temperature. An analog bar graph of measured values lets you view reading trends. Waterproof, IP57-rated tester floats in water.
Catalog No. 50-993-150