Learn More
iQ Biosciences Expanded Human CD3 T Cells, 50M cells/vial
Supplier:  iQ Biosciences IQBHU1T50E
Encompass_Preferred
High-quality, high-viability (> 90%) CD3+ T cells expanded from purified human T cells. Expanded T cells are validated for retention of functional activity such as proliferation, cytokine production, and cytolytic activity. Applications include a wide variety of safety assessment and functional assays. All orders come with an iQ Certificate of Analysis.
Catalog No. NC2855549
Explore available promotions