Learn More
Electron Microscopy Sciences ExP Vacuum Wand Kit for 12" (300mm) wafers, Molded PEEK wafer tip, ⅛" coiled hose and stand
SDP
Supplier:  Electron Microscopy Sciences 7193203
Encompass_Preferred
ExP Vacuum Wand wafer handling kits are a lower cost alternative to the Porta-Wand and Porta-Vac II systems, which share the same tips. Has a push-button to close the vacuum and release the wafer. The included stand keeps the button pressed when the wand is not in use. Comes complete with an ⅛" Inner Diameter coiled vacuum hose and a wafer tip. Must use a vacuum pump or vacuum line. Able to be used with our Vacuum Pick Up System. Comes with 1/8" coiled hose and stand.
Includes: Molded PEEK wafer tip, ⅛" coiled hose and stand
Catalog No. 50-193-1156