Learn More
EVOS™ EVOS™ Stage Plate for Automated Stage, one Nunc T-175 flask
EVOS™ vessel holders and stage plates fit a wide range of vessels, flasks, plates, dishes, and slides.
Supplier: EVOS™ AMEPVH042
Description
Product Features:- EVOS™ vessel holders allow a perfect fit of your microscope slide, cell culture flask or dish, or micro-well plate to the stage of the EVOS imaging systems for increased precision in sample alignment.
- EVOS stage plates hold vessel holders, thereby expanding the number of vessel holders available for the EVOS systems.
- With a snug and secure fit, yet readily switched, these accessories make imaging samples in a range of vessel holders convenient and easy.
Specifications
Imaging System Stage Plate | |
Stage Plate for Automated Stage, one Nunc™ T-175 flask | |
1 Nunc T-175 flask | |
Room Temperature |
EVOS | |
EVOS™ FL Auto Imaging System | |
259 mm x 204 mm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.