Learn More
EVOS™ EVOS™ Stage Plate Adapter
This adapter accommodates vessel holders AMEPVH001 through AMEPVH018 for precision in sample alignment
Supplier: EVOS™ AMEP4692
Description
The EVOS™ Stage Plate Adapter is designed to fit into the EVOS™ Staging Plate with 160 X 110 mm opening. . With a snug and secure fit, yet readily switched, the plate/adapter combination make imaging samples in a range of vessel holders convenient and easy.For additional choices, see the EVOS™ Guide to Vessel Holders and Stage Plates.
Explore the entire EVOS™ line of imaging systems and accessories.
Specifications
Stage Plate Adapter | |
EVOS™ FL Color Imaging System, EVOS™ XL Imaging System, EVOS™ FL Imaging System | |
1 Holder |
EVOS | |
Staging Plate |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only. Not for use in diagnostic procedures.