Learn More
Bullard™ EVAHL PAPR Set with PAPRFC4 HEPA/OV/AG Combination Filter
Offers cutting edge hazardous location blower unit with the state-of-the-art Sightline hood. The STLN2K offers unparalleled comfort with 320 degree unobstructed vision and tremendous overall comfort.
Supplier: Bullard™ EVAHLSTLN2KFC4
Description
Unique 320 degree vision to maximize vision and reduce claustrophobia; beveled bib to guarantee proper fit and simplify donning process; three channel air flow distribution and innovative drawstring / clip combination make the Sightline a great choice for all pharmaceutical operators.
- PAFLEXBR breathing tube contours to all body types and eliminates any bowing effect. Blower unit provides up to 8.5 CFM airflow.
- EVAHL blower features two speeds, dual alarms, and visual fuel gauge.
- Comfortable belt is easy to clean. Unique flex breathing tube contours to the operator body and reduces SKUs.
Specifications
Black, White | |
Combination Organic Vapor/Acid Gas/HEPA | |
PAPR Assembly with Hood | |
One Size |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.