Learn More
Bullard™ EVA PAPR Hood Kit with PAPRFC4 HEPA/OV/AG Combination Filter
Features the Sightline hood, offering 320 degree unobstructed vision and unparalleled comfort. This new hood is paired with the reliable EVA PAPR unit which provides up to 8.5 CFM airflow.
Supplier: Bullard™ EVASTLN2KFC4
Description
This kit features the EVA blower unit and new EVA PAPR hood, which has a two speed blower and can provide up to 8.5 CFM airflow to the operator.
- Dual alarms and visual fuel gauge are just some of the blower features.
- The belt is comfortable and easy to clean.
- Bullard's new Sightline hood sets the industry standard with maximum vision, beveled bibs, three point air distribution and innovative drawstring/clamp combination to maximize fit and comfort.
- The PAFLEXBT contours to operators body, and prevents and bowing effect.
Specifications
Black, White | |
Combination Organic Vapor/Acid Gas/HEPA | |
PAPR Assembly with Hood | |
One Size |