Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Ethyl Ether, UniSolv(R), 99.9%, Spectrum™ Chemical
Click to view available options
Quantity:
4 L
4 x 4 L
Packaging:
Amber Glass Bottle
Description
Spectrum™ Chemical Ethyl Ether, UniSolv(R) is also referred to as sulfuric ether, diethyl ether or even just simply as ether. This liquid was historically used as an anesthetic, but in more recent years been replaced by other more effective anesthetics. It has however some other uses such as fuel and solvent. It′s an organic compound that is extremely flammable and volatile on top of being colorless.
Specifications
CAS Max % | 100% |
UN Number | UN1155 |
Percent Purity | 99.9% |
Quantity | 4 x 4 L |
Packaging | Amber Glass Bottle |
Residue after Evaporation | 1 ppm |
Titratable Acid | 0.0002 meq/g |
Water | 0.02% |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction