Learn More
Med Vet International Ethicon Permahand, 5-0 Silk Suture, 30in. C-1, 36/BOX
Supplier: Med Vet International K890H
The Ethicon K890H suture is a high-quality non-absorbable Silk surgical suture. It measures 30in. in length, has a size of 5-0, and are dyed black to provide greater visibility while suturing. This suture comes with a C-1 taper point needle and has 36 sutures in a box.