Learn More
Med Vet International Ethicon Prolene Polypropylene Suture, Size 4-0, PS-2, 18", 12/Box, 8682G
Supplier: Med Vet International 8682G
The Ethicon 8682G suture is a high-quality non-absorbable Polypropylene surgical suture. It measures 18in. in length, has a size of 4-0, and are dyed black to provide greater visibility while suturing. This suture comes with a PS-2 Precision Point reverse cutting needle and has 12 sutures in a box.