Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Estradiol, Hemihydrate, Soy Base, Micronized, USP, 97-103%, Spectrum™ Chemical
Click to view available options
Quantity:
1 g
25 g
100 g
500 g
1 kg
Packaging:
Amber Glass Bottle
Poly Bottle
Description
Spectrum™ Chemical Estradiol, Hemihydrate, Soy Base, Micronized, USP
Specifications
CAS Max % | 100% |
Melting Range | 173°-179°C |
Packaging | Poly Bottle |
Quantity | 1 kg |
Specific Rotation | +76° to +83° |
Percent Purity | 97 to 103% |
Grade | USP |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction