Learn More
Medchemexpress LLC HY-15461 5mg Medchemexpress, Ertugliflozin CAS:1210344-57-2 Purity:>98%
Supplier: Medchemexpress LLC HY154615MG
Medchemexpress, HY-15461 5mg Ertugliflozin CAS:1210344-57-2 Ertugliflozin (PF-04971729) is a potent, selective and orally active inhibitor of the sodium-dependent glucose cotransporter 2 (SGLT2), with an IC50 of 0.877 nM for h-SGLT2. Has the potential for the treatment of type 2 diabetes mellitus[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.