Learn More
Impacto™ Protective Elbow Pad
Impacto™ ER402 Ergotech Elbow Pad protects and cushions the elbow
Supplier: Impacto™ ER40240
Description
Impacto™ ER402 Elbow Protectort
- Contoured closed cell foam padding protects elbow from cumulative trauma such as abrasion and bruising
- Cushions elbow against pressure point irritation and impact
- Pull-on style with durable rayon outer cover and moisture-wicking knit cotton liner
Specifications
Elbow Protector | |
Pull-on | |
Large | |
Suede |