Learn More
United Scientific™ Equipotential Trough Kit
Equipotential Trough Kit allows students to plot equipotential lines in an electric field between two electrodes held at different electric potentials.
Supplier: United Scientific™ UNEQPTK01
This item is not returnable.
View return policy
Description
- A simple setup keeps electrodes partially immersed in water containing an electrolyte, allowing a small current to flow between the electrodes
- Students can then sample the resulting electric field using the included multimeter
- Contents:
- Plastic trough, 33 x 25 cm, with printed orientation grid and fitted with three electrode support posts
- Wire harness with snap connector and two alligator clips
- 2 straight metal strip electrodes, 10 cm long
- 2 short rod electrodes
- 2 electrode support arms
- Sample point arm
- Digital multimeter
- Activity guide
Specifications
Physics |