Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Eppendorf™ twin.tec™ 96-Well Microbiology PCR Plates
Sterile, individually blistered, for all applications that need ultimate purity or certified absence of bacterial DNA
$180.50
Specifications
Certifications/Compliance | Human DNA, DNase, RNase-free |
---|---|
Format | Standard |
Array | 8 x 12 |
Autoclavable | Autoclavable |
Material | Polypropylene |
Catalog Number | Mfr. No. | Color | Volume (Metric) Well | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Volume (Metric) Well | Price | Quantity & Availability | ||||
E0030129300
|
Eppendorf™
0030129300 |
Clear | 150 μL |
Pack of 10 for $180.50
|
|
||||
E0030129318
|
Eppendorf™
0030129318 |
Blue | 150 μL |
Pack of 10 for $180.50
|
|
||||
E0030129326
|
Eppendorf™
0030129326 |
Clear | 250 μL |
Pack of 10 for $180.50
|
|
||||
E0030129334
|
Eppendorf™
0030129334 |
Blue | 250 μL |
Pack of 10 for $180.50
|
|
||||
Description
- Thin-walled for optimum heat transfer to the sample
- Solid and torque-resistant
- Improved well-to-well tolerance
- Minimal binding of DNA, RNA and enzymes
- Cut-away corner and alphanumeric grid referencing
- Raised well rims for effective sealing; also reduces risk of cross-contamination
- Autoclavable (121°C, 20 min.)
- Certified to be free of any detectable human DNA, DNase, RNase and PCR inhibitors†
Skirted Plates
- Compatible with automated systems
- Skirted for label or bar code
- Fits most thermal cyclers and the MegaBACE™ capillary sequencer
- Within SBS footprint recommendations (127.76 ×85.47mm ±0.25mm)
- Stackable
- Eight holes in the skirt aid plate positioning and removal from the thermal cycler block
- Low-profile design enhances efficiency of PCR and enables the highest efficiency for small sample volumes
- 150μL max. well volume (when used with cap strips)
- Ideal for quantitative real-time PCR
- Fits most thermal cyclers
- 250μL max. well volume (when used with cap strips)
Specifications
Human DNA, DNase, RNase-free | |
8 x 12 | |
Polypropylene | |
Sterile | |
10/Pk. | |
Clear |
Standard | |
Autoclavable | |
96 | |
RT-PCR applications | |
Yes |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title