Learn More
Eppendorf™ Maxipettor™ Maxitip™ Tips: Barrel/Pistons
Supplier: Eppendorf™ 022291408
Description
For Maxitip S™ tips. Kit includes 10 barrels with pistons, 100 Maxitip S tips, two Maxitip G™ tips and 10 valves.
Specifications
Barrel with Piston for Maxitip S Kit | |
Maxitip S |