Learn More
R Biopharm Inc ENZYTEC LIQUID D-GLUCOSE
Supplier: R Biopharm Inc E8140
The Enzytec™ Liquid product line enables the enzymatic determination of sugars, organic acids and other food components. These reagents are all liquid and ready-to-use, which brings a huge improvement in the lab organization:Test format Test-kit with 2 x 25 determinations for manual use,(500 tests on automated systems),2 x 50 ml R1 and 2 x 12.5 ml R2LOD (Detection Limit) 4.0 mg / LLOQ (Limit of quantification 10 mg / LDetected analyte D-GlucoseEvaluation 340 nm
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.