Learn More
Bioassay Systems EnzyChrom™ Kinase Assay Kit. Rapid homogenous assay for protein kinase activity and high-throughput screen for kinase inhibitors.
Supplier: Bioassay Systems EKIN400
Rapid homogenous assay for protein kinase activity and high-throughput screen for kinase inhibitors. Key Features: Safe. Non-radioactive assay. Sensitive and accurate. As low as 0.01 U/L kinase can be quantified. Homogeneous and convenient. "Mix-incubate-measure" type assay. The whole assay involves adding a single working reagent and incubation for 10 min at room temperature. Robust and amenable to HTS: Assay can tolerate up to 300 µM ATP and 10% dimethylsulfoxide (DMSO). Z factors of > 0.6 are routinely observed in 96/384-well plates. Can be readily automated on HTS liquid handling systems for tens of thousands of assays per day. Method: FL530/585nm. Samples: All kinases. Species: All. Procedure: Assay takes 10 min. Kit size: 400 tests. Detection limit: 0.01 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.