Learn More
Bioassay Systems EnzyChrom™ GSH/GSSG Assay Kit. For quantitative determination of reduced and oxidized glutathione (GSH/GSSG).
Supplier: Bioassay Systems EGTT100
For quantitative determination of reduced/oxidized glutathione (GSH/GSSG) and evaluation of drug effects on their metabolism. Key Features: Sensitive and accurate. Linear detection range 0.01-3 µM GSH equivalents with a detection limit of 10 nM GSH equivalents. Method: OD412nm. Samples: Whole blood, plasma, serum, urine, tissue and cell extracts. Species: All. Procedure: Assay takes 20 min. Kit size: 100 tests. Detection limit: 10 nM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.