Learn More
Bioassay Systems EnzyChrom™ Glycerol Assay Kit. For quantitative determination of glycerol and evaluation of drug effects on glycerol metabolism.
Supplier: Bioassay Systems EGLY200
For quantitative determination of glycerol and evaluation of drug effects on glycerol metabolism. Key Features: Sensitive and accurate. Use as little as 10 µL samples. Linear detection range in 96-well plate: 10 to 1000 µM (92 µg/dL to 9.2 mg/dL) glycerol for colorimetric assays and 2 to 50 µM for fluorimetric assays. Simple and convenient. The procedure involves addition of a single working reagent and incubation for 20 min at room temperature, compatible for HTS assays. Improved reagent stability. The optimized formulation has greatly enhanced the reagent and signal stability. Method: OD570nm, or FL530/585nm. Samples: Biological, food, beverage etc. Species: All. Procedure: Assay takes 20 min. Kit size: 200 tests. Detection limit: 2 µM.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.