Learn More
Bioassay Systems EnzyChrom™ Cholesterol Assay Kit. For quantitative determination of cholesterol and evaluation of drug effects on cholesterol metabolism.
Supplier: Bioassay Systems ECCH100
For quantitative determination of cholesterol and evaluation of drug effects on cholesterol metabolism. Key Features: Sensitive and accurate. Detection limit of 5 mg/dL, linearity up to 300 mg/dL cholesterol in 96-well plate assay. Convenient. Room temperature assay. No 37°C heater is needed. High-throughput. Can be readily automated as a high-throughput 96-well plate assay for thousands of samples per day. Method: OD340nm. Samples: Serum, plasma etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 5 mg/dL.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.