Learn More
Bioassay Systems EnzyChrom™ Catalase Assay Kit. For quantitative determination of catalase activity and evaluation of drug effects on catalase activity.
Supplier: Bioassay Systems ECAT100
For quantitative determination of catalase activity and evaluation of drug effects on catalase activity. Key Features: Sensitive and accurate. Use 10 µL sample. Linear detection range 0.2 to 5 U/L catalase activity. Simple and Convenient. The procedure involves adding a Substrate to sample, incubation for 30 min, followed by a Detection Reagent and reading the optical density or fluorescence intensity. Method: OD570nm, or FL530/585nm. Samples: Serum, plasma, urine, saliva, cell culture etc. Species: All. Procedure: Assay takes 40 min. Kit size: 100 tests. Detection limit: 0.2 U/L.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.