Learn More
Bioassay Systems EnzyChrom™ Adipolysis Assay Kit. For quantitative assay of adipolysis and evaluation of drug effects on adipolysis.
Supplier: Bioassay Systems EAPL200
For quantitative assay of adipolysis and evaluation of drug effects on adipolysis. Key Features: Sensitive and accurate. Use as little as 10 µL samples. Linear detection range in 96-well plate: 0.92 to 100 µg/mL (10 to 1000 µM) glycerol for colorimetric assays and 0.2 to 5 µg/mL for fluorimetric assays. Rapid and convenient.The procedure involves addition of a single working reagent and incubation for 20 min at room temperature. Robust and amenable to HTS assays. Potential interference by testing drugs is greatly reduced at 570nm. Compatible with culture media containing phenol red. Assays can be performed in 96 or 384-well plates. Method: OD570nm, or FL530/585nm. Samples: Cell culture media. Species: All. Procedure: Assay takes 20 min. Kit size: 200 tests. Detection limit: 0.92 µg/mL.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.