Learn More
3M™ Petrifilm™ Environmental Listeria Plate, Sold by AquaPhoenix Scientific

Detect the presence of Listeria in environmental samples. A sample ready culture medium containing selective agents, nutrients, a cold water soluble gelling agent and a chromogenic indicator.
$476.00 - $1454.00
Specifications
Content And Storage | Temperatures ≤ 8°C |
---|---|
Detects | Listeria |
Product Type | Identification Test |
Test Type | Environmental Test |
For Use With (Application) | Food processors and contract labs |
Description
- Proven cost effective tool for detection of environmental Listeria
- Provide accurate, efficient, consistent results for food processors and contract labs
- Unique results (qualitative, semi-quantitative and quantitative) allow identification of hot spots in plant, track progress against Listeria over time
- Rapid results in 26 to 30 hours from sample collection
- This plate has been awarded Performance Tested Method (PTM) #030601 for stainless steel, ceramic tile and sealed concrete
Specifications
Temperatures ≤ 8°C | |
Identification Test | |
Food processors and contract labs |
Listeria | |
Environmental Test |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.