Learn More
Perkin Elmer US LLC Environmental Hydrocarbons HATR Application Pack
Supplier:  Perkin Elmer US LLC L1608008
Encompass_Preferred
This Environmental Hydrocarbons Application Pack contains: HATR Accessory for the Spectrum Two with a 45° ZnSe flat top plate Environmental Hydrocarbons Touch Apps Software Kit The Touch Apps Software Kit includes the software and SOPs to support routine measurement of oil and grease in water and soil samples by FT-IR spectroscopy. Spectrum Touch software provides a simplified, workflow-driven operating environment; for faster, more reproducible analyses.
Catalog No. 50-211-7236