Learn More
Environmental Express™ Filters
Ideal for air particle monitoring and environmental testing
Supplier: Environmental Express™ F370800
Description
- Laser cutting ensures a clean, fiber-free filter edge
- Excellent temperature and chemical resistance
Specifications
37 mm |