Learn More
H-B Instrument™ Enviro-Safe™ General Purpose Liquid-In-Glass Thermometers

Certified environmentally safe thermometers replace dangerous mercury.
Supplier: H-B Instrument™ B605020100
Description
- Earth friendly, EnviroKleen™ certified thermometers
- Feature green, non-toxic, biodegradable, Enviro-Safe™ liquid against lead-free, white back glass
- Meet government, education, and industry mandates to purchase “Green” products
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Order Info
Should you require an individualized Thermometer Calibration Report, factory calibrated thermometers or custom calibration services are available.
Certifications
Individually serialized thermometers are traceable to NIST; include a multi-language Statement of Accuracy and Traceability.
Compliance
Manufactured and tested by USA quality team.
Specifications
ISO/IEC 17025:2005, A2LA, NIST, DAkkS/PTB | |
Partial | |
11.8 in. | |
Glass | |
110°C | |
-20°C to +110°C | |
1°C |
Chemical laboratories, universities, scientific institutions, food and beverage processing, petrochemical laboratories, pharmaceutical industry, medical industry, manufacturers, dairies, breweries and distilleries | |
Multilanguage instructions, statement of accuracy | |
300 mm | |
Enviro-Safe™ Liquid | |
−20°C | |
±1°C | |
76 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.