Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mopec Envelope Zipper, Heavy Duty Body Bag
Polyethylene body bag with reinforced nylon
Supplier: Mopec BE165WHT
Description
- Zipper style: Envelope Style, rust resistant zipper
- Handles: No
- Shelf life: 10 years
- Recommended for Cremation
Specifications
Heavy Duty | |
Polyethylene, Reinforced Nylon | |
90 x 40 in. |
White | |
8 mil | |
1000 lb. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction