Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Liofilchem™ EnteroPluri-Test
EnteroPluri-Test is a 12-sector system containing special culture media for the identification of Enterobacteriaceae and other Gram-negative, oxidase negative bacteria.
$234.47 - $532.88
Specifications
Disposable | Single-use |
---|---|
For Use With (Application) | Identification of Gram-negative or Oxidase Negative Bacteria |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
22-778-080
|
Liofilchem™
78618U |
10 Pk. |
Pack of 10 for $234.47
|
|
|||||
22-778-079
|
Liofilchem™
78619U |
25 Pk. |
Pack of 25 for $532.88
|
|
|||||
Description
- Allows the simultaneous inoculation of all media present in the sectors and the execution of 15 biochemical reactions
- Microorganism is identified evaluating the color change of different culture media after 18 to 24 hr incubation at 36 ±1°C and by code number obtained from interpretation of the biochemical reactions
- Used for identification of Gram-negative, oxidase negative bacteria isolated on selective culture media for Enterobacteriaceae, such as: MacConkey Agar (MCA), Eosin Methylene Blue Agar (EMBA), Salmonella and Shigella Agar (SSA), Hektoen Enteric Agar (HEEA), or non-selective culture media
- Tests come individually packed
Specifications
Single-use | |
Identification of Gram-negative or Oxidase Negative Bacteria |
Non-hazardous' according to current legislation, but falls within specific field of application where a safety data sheet must be supplied, as product can cause phenomena of sensitization in sensitive subjects if in contact with skin.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title