Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
3M™ Enrichment Pouch, Sold by AquaPhoenix Scientific
Heavyweight, puncture-resistant pouch, ideal for many sample preparation and enrichment procedures
$155.23
Specifications
Certifications/Compliance | ISO 9001:2008 |
---|---|
Media Type | 225 mL buffered peptone water broth |
Product Type | Enrichment |
Test Type | Environmental Test |
For Use With (Application) | Sample preparation and enrichment procedures |
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Includes | Price | Quantity & Availability | |||||
S26747
|
3M™
QEBPW225F |
Filter |
Each for $155.23
|
|
|||||
S26746
|
3M™
QEBPW225 |
No filter | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Gamma irradiated, ready-to-use pouch with 225 mL buffered peptone water broth requires no mixing or sterilizing
- Flat bottom, stand-up pouch ensures stability when adding samples
- Wire tie holds top open for easy sample addition and closes pouch for transportation
- Multi-laminate polyester pouch is puncture resistant
- 2 year shelf life from date of manufacture provides long-term readiness
Specifications
ISO 9001:2008 | |
Enrichment | |
Sample preparation and enrichment procedures |
225 mL buffered peptone water broth | |
Environmental Test | |
32/Cs. |
Safety and Handling
ShelfLife : 2 years
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title