Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ENG Scientific Urinary Sediment Stain
A quick staining procedure by which a drop of stain solution is directly added to urine sediment.
$449.00
Specifications
Type | Urinary Sediment Stain |
---|---|
For Use With (Application) | Visualize neutrophils, glitter and squamous cells, bacteria and hyaline, granular and cellular casts |
Includes | Crystal violet, ammonium oxalate, ethanol, safranin |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
ES3150
|
ENG Scientific
3150 |
2 x 100 mL |
Pack of 2 for $449.00
|
|
|||||
ES3151
|
ENG Scientific
3151 |
1 x 100 mL | Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please Sign In to view product availability in your area. | N/A | |||||
Description
- Modification of Sternheimer and Malbin
- Contents: Phloxine, Propylene Glycol, Sodium Carbonate Phloxine, Propylene Glycol, Sodium Carbonate
- Solution made from certified dyes
Specifications
Urinary Sediment Stain | |
Visualize neutrophils, glitter and squamous cells, bacteria and hyaline, granular and cellular casts | |
Crystal violet, ammonium oxalate, ethanol, safranin |
For in vitro diagnostic use only.
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title