Learn More
BioVendor, LLC ENDOTRAP HD5,5ML SETTLE RESIN
SDP
Supplier:  BioVendor, LLC LET0023
Encompass
TypeEndoTrapHDDescriptionEndoTrap HD removes endotoxins (lipopolysaccharides/LPS) from aqueous solutions of a wide range of samples such as proteins, antibodies, vaccines, nucleic acids, buffers and cell extracts with excellent sample recovery rate. The EndoTrap HD resin is especially designed to meet the needs of pharma and biotech industry for the efficient removal of endotoxins in biomanufacturing processes as well as any application that requires highest endotoxin removal rates.EndoTrap HD fulfils all requirements for industrial applications, and it can easily be integrated in/at early or late-stage purification processes. Moreover, High-Definition Matrix is also available for small-scale purification as pre-filled columns. EndoTrap HD is manufactured by LIONEX GmbH in Germany.
Catalog No. NC1668923