Learn More
World Precision Instrument Endoscope Sterilization Tray with Lens Basket
Supplier: World Precision Instrument WP4046
- Safely sterilize and store expensive endoscopes
- Silicone pads to protect endoscope during handling and storage
- Small lens basket included
- Locking clip
- 640 x 150 x 77 mm