Learn More
World Precision Instrument Endoscope Sterilization Baskets-WP-4043
Supplier: World Precision Instrument WP4043
- Safely sterilize and store expensive endoscopes
- Silicone pads to protect endoscope during handling and storage
- Electropolished
- Burr free to prevent injury
- Locking clip
- Variety of sizes