Learn More
World Precision Instrument Endoscope Sterilization Tray, Short
Supplier: World Precision Instrument WP4054
- Safely sterilize and store expensive endoscopes
- Silicone pads to protect endoscope during handling and storage
- Locking clip
- 310 x 150 x 77 mm
This item is not returnable.
View return policy