Learn More
Cole-Parmer Endecotts MINOR 200 Compact Electromagnetic Sieve Shaker; 115 VAC, 60 Hz
Supplier: Cole-Parmer 5888670
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Sift heavy minerals, light organic samples, and fine pharmaceutical powders with this electromagnetic shaker. It has no motor and no moving parts—making it virtually maintenance free. The shaker provides a unique vibratory motion by using a horizontal and vertical sieving action to effectively separate materials. Use the built-in 60-minute timer to easily repeat any test. Shaker holds up to 8 full-height or 16 half-height, 8'' diameter sieves with bottom pan and cover. Sieves are held securely in place with clamps.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.