Learn More
Thermo Scientific EMT DUO TORCH
Supplier:  Thermo Scientific 842312051841
Encompass
Duo Torch (EMT); ICP quartz torch for use with iCAP 6000 Series Duo instruments using; the EMT Torch holder.
Catalog No. NC9712332
Explore available promotions