Learn More
Electron Microscopy Sciences EMS Formvar/Silicon Monoxide Film on 300 mesh Nickel Grid
Supplier: Electron Microscopy Sciences FSF300NI
A formvar film stabilized with a thin film of Silicone monoxide. Silicone monoxide produces a desirable support film because it offers low background contrast and it is stable under the electron beam and it is more hydrophilic than carbon film. All of our coated grids are optically checked followed by batch testing in the EM. Packed in grid storage box. NOTE: All of our film is laid on the shiny side of the grid.
Grid: Nickel
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.