Learn More
Electron Microscopy Sciences EMS 50 Grid Storage Box
Supplier: Electron Microscopy Sciences 7114601
Used for the storage of TEM grids for routine grid handling, transport and long term TEM grid storage. For standard grids that are 3.05 mm in diameter. Anti Static treated