Learn More
Seca Corp 264 Digital wall-mounted stadiometer with backlit display on headpiece
Supplier: Seca Corp 2641900009
The goal of accomplishing more in fewer steps is as important in medicine. seca 264 helps make that possible. For example, the heel positioner and the seca Frankfurt Line on the headpiece ensure that the patient is in the right position and thereby contribute to obtaining accurate measurements. Height measurements can be read with a quick glance at the white backlit display on the headpiece. • Digitally measures both children and adults • Frankfurt Line for precise head alignment • Backlit LCD display in headpiece • Non-slip rubber mat with heel-positioner • Measuring range: 11 in – 7 ft 2 in / 30 – 220 cm • Graduation: 1/8 in / 1 mm • Functions: HOLD, inch/cm switch-over, user-defined zero setting, automatic switch-off
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.