Learn More
CDS Analytical™ Empore™ SDB-RPS StageTips
SDP
SDB-RPS (styrenedivinylbenzene reverse phase sulfonate) polymer sorbent StageTips are SPE micropipette tips for the desalting of large peptides and proteins, as well as peptide fractionation.
Supplier:  CDS Analytical™ 70201910049EA
 View more versions of this product
Catalog No. 13-110-061
Add to Cart
This item is not returnable. View return policy
Description
Description
The CDS™ Empore™ StageTips SDB-RPS (styrenedivinyl benzene reverse phase sulfonate) polymer sorbent solid phase extraction micropipette tips have two layers for increased capacity.
  • Combines good retention capacity with high recoveries
  • Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Specifications
12 μm
DVB (Polystyrene-divinylbenzene)
96 Pk.
Empore
200 μL
Product Suggestions
Product Suggestions