Learn More
CDS Analytical™ Empore™ C18/SCX/C18 StageTips
SDP
StageTips with C18/SCX/C18 layers, C18 and cation exchange (SCX) SPE micropipette tips are for the desalting of large peptides and proteins, as well as peptide fractionation.
Supplier:  CDS Analytical™ 70201910088
 View more versions of this product
Catalog No. 13-110-070
Add to Cart
This item is not returnable. View return policy
Description
Description
CDS™ Empore™ StageTips with C18/SCX/C18 layers, C18 and cation exchange (SCX) solid phase extraction micropipette tips.
  • Combines good retention capacity with high recoveries
  • Recommended for desalting of large peptides and proteins and peptide fractionation at acidic, neutral, and basic pH conditions
Specifications
Specifications
12 μm
DVB (Polystyrene-divinylbenzene)
960 Cs.
Empore
200 μL
Product Suggestions
Product Suggestions