Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Dukal™ Emojis Bandages
Sterile adhesive bandages, size 3/4 x 3 in.
Supplier: Dukal™ 15606
Description
- Absorbent, non-stick pad for easy removal
- Unique, long-lasting adhesive helps bandage stay on, to keep cuts clean
- Multi-purpose family protection for cuts and scrapes
- Comes with the preferred and patented Stat Strip™; easy opening wrapper that nurses request
Specifications
Adhesive Bandage | |
3 in. | |
100 Pk. | |
Box | |
Emojis |
Sterile | |
3/4 in. | |
1.9 x 7.6 cm | |
Keeps cuts clean |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction