Learn More
ZORO TOOLS EMF METER LCD INCLUDES (3) AA
Supplier: ZORO TOOLS 132823
EMF Meter, LCD, Include (3) AAA Batteries.Multi-Field EMF Meter. Ranges: 2000mG Magnetic, 2000V/m Electric, 554.6mW/m2 RF. Frequency Range (Hz): 50 MHz to 3.5 GHz RF, 50 to 60 Hz Electric, 50 to 60 Hz Magneti
This item is not returnable.
View return policy