Learn More
Sklar Instruments Emesis Basin, Sklar, Stainless Steel. Sklar Emesis Bowls, also known as a kidney dish due to their shape. This bowl measures 6-5/8 in. x 3-1/2 in. x 1-1/2 in. and hold up to 10 oz. of fluid. EA/1.
SDP
Supplier:  Sklar Instruments 101497
Encompass_Preferred
Emesis Basin, Sklar, Stainless Steel. Sklar Emesis Bowls, also known as a kidney dish due to their shape, are used in surgical settings for collecting fluids or debris that may exit a patients body during a procedure. The shape of the bowl allows for it to be placed up closely against the patient. It may also be used as a collecting container for soiled dressings until they can be properly disposed. This bowl measures 6-5/8 in. x 3-1/2 in. x 1-1/2 in. and hold up to 10 oz. of fluid.
Catalog No. 50-143-7007
Explore available promotions