Learn More
MilliporeSigma™ Vent Filters for Pure Water Storage Reservoirs
For use with Pure Water Reservoirs
$210.00 - $436.00
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | For Use With (Equipment) | Price | Quantity & Availability | |||||
TANKMPK02
|
MilliporeSigma™
TANKMPK02 |
RiOs™ 5/8/16 system |
Pack for $210.00
|
|
|||||
TANKMPK01
|
MilliporeSigma™
TANKMPK01 |
Elix™ 3/5/10 system |
Each for $436.00
|
|
|||||
Description
For use with Pure Water Reservoirs
- Polyethylene vent filters protect purified water stored in Pure Water Reservoirs when used with low-volume Elix™ or RiOs™ Water Purification Systems from ingress of airborne microorganisms
Specifications
Polyethylene | |
Vent Filter |