Learn More
MilliporeSigma™ UV Lamps and Assemblies for Water Purification Systems
Control bacteria and oxidize organics
Supplier: MilliporeSigma™ QUVLQSL01
Description
Control bacteria and oxidize organics
- Replacement parts for EMD Millipore Water Purification Systems and A10 TOC Monitor
Specifications
UV Lamp | |
Milli-Q™ gradient and synthesis model (blue door) water purification system | |
254/185 nm |