Learn More
MilliporeSigma™ Upstate ChIPAb+ Trimethyl-Histone H4 (Lys20) - ChIP Validated Antibody and Primer Set
Trimethyl histone H4 (Lys20) antibody against synthetic peptide corresponding to amino acids 18-22 of histone H4 trimethylated on lysine 20
Supplier: MilliporeSigma™ 17671
Includes: Negative control rabbit antibody and control primers specific for human beta-globin promoter
Specifications
HIST2H4, H4/A, H4/B, H4/C, H4/D, H4/E, H4/G, H4/H, H4/I, H4/J, H4/M, H4/N, H4/O, H4/a, H4F2, H4FA, H4FB, H4FC, H4FD, H4FE, H4FG, H4FH, H4FI, H4FJ, H4FK, H4FM, H4FN, H4FO | |
ChIPAb+ Trimethyl-Histone H4 (Lys20) - ChIP Validated Antibody and Primer Set |